missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-methylpropiophenone 96.0+%, TCI America™

Catalog No. p-7130848
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H099125G 25 g
H0991100G 100 g
H0991500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H099125G Supplier TCI America Supplier No. H099125G
Only null left

Chemical Identifiers

CAS 7473-98-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00040710
InChI Key XMLYCEVDHLAQEL-UHFFFAOYSA-N
Synonym 2-Benzoyl-2-propanol
PubChem CID 81984
IUPAC Name 2-hydroxy-2-methyl-1-phenylpropan-1-one
SMILES CC(C)(C(=O)C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-2-methylpropiophenone
EINECSNumber (3)-3581
RTECSNumber US2977245
TSCA Yes