missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-methylbutyric Acid 98.0+%, TCI America™

Catalog No. H06365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06365G 5 g
H063625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06365G Supplier TCI America Supplier No. H06365G
Only null left

Chemical Identifiers

CAS 3739-30-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00004476
InChI Key MBIQENSCDNJOIY-UHFFFAOYNA-N
PubChem CID 95433
ChEBI CHEBI:68454
IUPAC Name 2-hydroxy-2-methylbutanoic acid
SMILES CCC(C)(O)C(O)=O

Specifications

Melting Point 72°C
Color White
Quantity 5 g
Formula Weight 118.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-2-methylbutyric Acid
EINECSNumber (2)-1429
TSCA No