missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-azaadamantane 98.0+%, TCI America™

Catalog No. h1404200mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
5 g
Catalog No. Quantity
H1404200MG 200 mg
H14041G 1 g
H14045G 5 g
3 options

Catalog No. H1404200MG

Supplier: TCI America H1404200MG

Only null left

Chemical Identifiers

CAS 1155843-79-0
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.225
InChI Key SFXINVPZIZMFHM-UHFFFAOYSA-N
Synonym AZADOL(regR)
PubChem CID 51037224
SMILES C1C2CC3CC1CC(C2)N3O

Specifications

Quantity 200 mg
Formula Weight 153.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-2-azaadamantane