missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-azaadamantane 98,0+%, TCI America™

Numéro de catalogue. H14045G
Modifier l'affichage
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H14045G 5 g
H1404200MG 200 mg
H14041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. H14045G Fournisseur TCI America Code fournisseur H14045G

Identifiants chimiques

CAS 1155843-79-0
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.225
InChI Key SFXINVPZIZMFHM-UHFFFAOYSA-N
Synonym AZADOL(regR)
PubChem CID 51037224
SMILES C1C2CC3CC1CC(C2)N3O

Spécifications

Quantity 5 g
Formula Weight 153.23
Percent Purity ≥98.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material 2-Hydroxy-2-azaadamantane