missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrogen 1-Methyl (1S,2R)-1,2,3,6-Tetrahydrophthalate 98.0+%, TCI America™

Catalog No. H1652500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1652500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1652500MG Supplier TCI America Supplier No. H1652500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88335-93-7
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.19
MDL Number MFCD00757425,MFCD00075490
InChI Key MYYLMIDEMAPSGH-UHFFFAOYNA-N
Synonym 1s,2r-1-methyl cis-1,2,3,6-tetrahydrophthalate, 1r,6s-6-methoxycarbonyl cyclohex-3-enecarboxylic acid, 1r,6s-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid, 1r,6s-6-methoxycarbonyl cyclohex-3-ene-1-carboxylic acid, 1s,2r-1-methyl cis-4-cyclohexene-1,2-dicarboxylate, 1r,6s-6-methoxycarbonyl-1-cyclohex-3-enecarboxylic acid, 1s,2r-1-methylcis-1,2,3,6-tetrahydrophthalate, 2-hydrogen 1-methyl 1s,2r-1,2,3,6-tetrahydrophthalate, 1 r,6s-6-methoxycarbonyl-3-cyclohexene-1-carboxylic acid, 1r, 2s-2-methoxycarbonylcyclohex-4-ene-1-carboxylic acid
PubChem CID 5325907
IUPAC Name 6-(methoxycarbonyl)cyclohex-3-ene-1-carboxylic acid
SMILES COC(=O)C1CC=CCC1C(O)=O

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 500 mg
Formula Weight 184.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrogen 1-Methyl (1S,2R)-1,2,3,6-Tetrahydrophthalate