missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinoquinoline 98.0+%, TCI America™

Catalog No. H01781G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01781G 1 g
H01785G 5 g
H017825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H01781G Supplier TCI America Supplier No. H01781G

Chemical Identifiers

CAS 15793-77-8
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00041860
InChI Key QMVCLSHKMIGEFN-UHFFFAOYSA-N
PubChem CID 85111
IUPAC Name 2-hydrazinylquinoline
SMILES NNC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 142°C
Quantity 1 g
Synonym 2-hydrazinoquinoline, 2-hydrazinylquinoline, 2-quinolylhydrazine, 2 1h-quinolinone, hydrazone, quinoline, 2-hydrazino, 2-hydrazinequinoline, quinoline, 2-hydrazinyl, quinoline,2-hydrazinyl, .alpha.-quinolylhydrazine, alpha-quinolylhydrazine
Formula Weight 159.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrazinoquinoline