Learn More
2-Hydrazinoquinoline, 97%
CAS: 15793-77-8 | C9H9N3 | 159.19 g/mol
Supplier: Thermo Scientific Chemicals H5070003
Description
2-Hydrazinoquinoline acts as as a derivatization agent for LC-MS-based metabolomic investigation of diabetic ketoacidosis. It is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydrazinoquinoline acts as as a derivatization agent for LC-MS-based metabolomic investigation of diabetic ketoacidosis. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in methanol.
Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
Specifications
| 2-Hydrazinoquinoline | |
| 141°C to 143°C | |
| MFCD00041860 | |
| 2-hydrazinoquinoline, 2-hydrazinylquinoline, 2-quinolylhydrazine, 2 1h-quinolinone, hydrazone, quinoline, 2-hydrazino, 2-hydrazinequinoline, quinoline, 2-hydrazinyl, quinoline,2-hydrazinyl, .alpha.-quinolylhydrazine, alpha-quinolylhydrazine | |
| QMVCLSHKMIGEFN-UHFFFAOYSA-N | |
| quinolin-2-ylhydrazine | |
| 85111 | |
| 97% |
| 15793-77-8 | |
| C9H9N3 | |
| 1 g | |
| Soluble in methanol. | |
| NNC1=CC=C2C=CC=CC2=N1 | |
| 159.19 | |
| 159.19 |
Chemical Identifiers
| 15793-77-8 | |
| 159.19 | |
| QMVCLSHKMIGEFN-UHFFFAOYSA-N | |
| quinolin-2-ylhydrazine |
| C9H9N3 | |
| MFCD00041860 | |
| 85111 | |
| NNC1=CC=C2C=CC=CC2=N1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H315-H319-H335
EINECSNumber : 239-899-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only