missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinopyridine 98.0+%, TCI America™

Catalog No. p-7130785
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H088810G 10 g
H088825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H088810G Supplier TCI America Supplier No. H088810G
Only null left

Chemical Identifiers

CAS 4930-98-7
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006249
InChI Key NWELCUKYUCBVKK-UHFFFAOYSA-N
Synonym 2-hydrazinylpyridine, 2-hydrazinopyridine, 2-pyridylhydrazine, 2 1h-pyridinone, hydrazone, hydrazine, 2-pyridinyl, pyridine, 2-hydrazinyl, 2-pyridylhydrazone, pyridin-2-yl-hydrazine, pyridine, 2-hydrazino, pyridin-2-yl hydrazine
PubChem CID 78645
IUPAC Name 2-hydrazinylpyridine
SMILES NNC1=CC=CC=N1

Specifications

Melting Point 47°C
Boiling Point 185°C
Quantity 10 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrazinopyridine