missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydrazinobenzothiazole 99.0+%, TCI America™
$98.79 - $264.70
Chemical Identifiers
| CAS | 615-21-4 |
|---|---|
| Molecular Formula | C7H7N3S |
| Molecular Weight (g/mol) | 165.214 |
| MDL Number | MFCD00041849 |
| InChI Key | JYSUYJCLUODSLN-UHFFFAOYSA-N |
| Synonym | 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine |
| PubChem CID | 11988 |
| IUPAC Name | 1,3-benzothiazol-2-ylhydrazine |
| SMILES | C1=CC=C2C(=C1)N=C(S2)NN |
Chemical Identifiers
| 615-21-4 | |
| 165.214 | |
| JYSUYJCLUODSLN-UHFFFAOYSA-N | |
| 11988 | |
| C1=CC=C2C(=C1)N=C(S2)NN |
| C7H7N3S | |
| MFCD00041849 | |
| 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine | |
| 1,3-benzothiazol-2-ylhydrazine |
Specifications
| 615-21-4 | |
| C7H7N3S | |
| 5 g | |
| 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine | |
| C1=CC=C2C(=C1)N=C(S2)NN | |
| 165.214 | |
| 165.21 | |
| Crystalline Powder |
| 202°C | |
| MFCD00041849 | |
| 2811 | |
| JYSUYJCLUODSLN-UHFFFAOYSA-N | |
| 1,3-benzothiazol-2-ylhydrazine | |
| 11988 | |
| ≥99.0% (T) | |
| 2-Hydrazinobenzothiazole |
Safety and Handling
RTECSNumber : DL4725000
TSCA : Yes