missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinobenzothiazole 99.0+%, TCI America™

Catalog No. H01775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01775G 5 g
H017725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01775G Supplier TCI America Supplier No. H01775G

Chemical Identifiers

CAS 615-21-4
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.214
MDL Number MFCD00041849
InChI Key JYSUYJCLUODSLN-UHFFFAOYSA-N
Synonym 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine
PubChem CID 11988
IUPAC Name 1,3-benzothiazol-2-ylhydrazine
SMILES C1=CC=C2C(=C1)N=C(S2)NN

Specifications

Melting Point 202°C
Quantity 5 g
UN Number 2811
Formula Weight 165.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrazinobenzothiazole
RTECSNumber DL4725000
TSCA Yes