missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinobenzothiazole, 97%

Catalog No. AAA1800618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1800618 50 g
AAA1800609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1800618 Supplier Thermo Scientific Chemicals Supplier No. A1800618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 615-21-4 | C7H7N3S | 165.214 g/mol

It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.

Solubility
Solubility in hot methanol almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 615-21-4
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.214
MDL Number MFCD00041849
InChI Key JYSUYJCLUODSLN-UHFFFAOYSA-N
Synonym 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine
PubChem CID 11988
IUPAC Name 1,3-benzothiazol-2-ylhydrazine
SMILES C1=CC=C2C(=C1)N=C(S2)NN

Specifications

Melting Point 198°C to 202°C
Quantity 50 g
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 2-Hydrazinobenzothiazole
Hazard Category H302-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-416-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only