missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazino-4-(trifluoromethyl)pyrimidine 98.0+%, TCI America™

Catalog No. H15741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15741G 1 g
H15745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15741G Supplier TCI America Supplier No. H15741G

Chemical Identifiers

CAS 197305-97-8
Molecular Formula C5H5F3N4
Molecular Weight (g/mol) 178.118
MDL Number MFCD00115112
InChI Key XQCAINVUBSAUBJ-UHFFFAOYSA-N
Synonym 2-hydrazino-4-trifluoromethyl pyrimidine, 2-hydrazinyl-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ylhydrazine, n-4-trifluoromethyl pyrimidin-2-yl hydrazine, 4-trifluoromethyl pyrimidin-2-yl hydrazine, 1-4-trifluoromethyl pyrimidin-2-yl hydrazine, 2 1h-pyrimidinone, 4-trifluoromethyl-, hydrazone, buttpark 27\08-68, ksc498g6p, 2-hydrazino-4-trifluoromethylpyrimidine
PubChem CID 1268079
IUPAC Name [4-(trifluoromethyl)pyrimidin-2-yl]hydrazine
SMILES C1=CN=C(N=C1C(F)(F)F)NN

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 178.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrazino-4-(trifluoromethyl)pyrimidine