missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%

Catalog No. AAH6128703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6128703 1 g
AAH6128706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6128703 Supplier Thermo Scientific Chemicals Supplier No. H6128703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 197305-97-8 | C5H5F3N4 | 178.118 g/mol

2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

Solubility
Soluble in aqueous acid.

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 197305-97-8
Molecular Formula C5H5F3N4
Molecular Weight (g/mol) 178.118
MDL Number MFCD00115112
InChI Key XQCAINVUBSAUBJ-UHFFFAOYSA-N
Synonym 2-hydrazino-4-trifluoromethyl pyrimidine, 2-hydrazinyl-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ylhydrazine, n-4-trifluoromethyl pyrimidin-2-yl hydrazine, 4-trifluoromethyl pyrimidin-2-yl hydrazine, 1-4-trifluoromethyl pyrimidin-2-yl hydrazine, 2 1h-pyrimidinone, 4-trifluoromethyl-, hydrazone, buttpark 27\08-68, ksc498g6p, 2-hydrazino-4-trifluoromethylpyrimidine
PubChem CID 1268079
IUPAC Name [4-(trifluoromethyl)pyrimidin-2-yl]hydrazine
SMILES C1=CN=C(N=C1C(F)(F)F)NN

Specifications

Melting Point 99°C to 101°C
Quantity 1 g
UN Number UN2811
Solubility Information Soluble in aqueous acid.
Formula Weight 178.12
Percent Purity 97%
Chemical Name or Material 2-Hydrazino-4-(trifluoromethyl)pyrimidine
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only