missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexanoylfuran, 99%
CAS: 14360-50-0 | C10H14O2 | 166.22 g/mol
$94.97 - $308.93
Chemical Identifiers
| CAS | 14360-50-0 |
|---|---|
| Molecular Formula | C10H14O2 |
| Molecular Weight (g/mol) | 166.22 |
| MDL Number | MFCD00037354 |
| InChI Key | YUAYWSBSIJVIBS-UHFFFAOYSA-N |
| Synonym | 2-hexanoylfuran, 2-furyl pentyl ketone, 1-hexanone, 1-2-furanyl, pentyl 2-furyl ketone, 1-2-furyl-1-hexanone, 1-furan-2-yl hexan-1-one, 1-2-furyl hexanone, 2-furyl n-pentyl ketone, 1-2-furanyl-1-hexanone, furan, 2-hexanoyl |
| PubChem CID | 61738 |
| IUPAC Name | 1-(furan-2-yl)hexan-1-one |
| SMILES | CCCCCC(=O)C1=CC=CO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2230606
|
Thermo Scientific Chemicals
B2230606 |
5 g |
Each for $94.97
|
|
|||||
|
AAB2230614
|
Thermo Scientific Chemicals
B2230614 |
25 g |
Each for $308.93
|
|
|||||
Description
2-Hexanoylfuran is used as a food additives
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hexanoylfuran is used as a food additives
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 14360-50-0 | |
| 166.22 | |
| YUAYWSBSIJVIBS-UHFFFAOYSA-N | |
| 61738 | |
| CCCCCC(=O)C1=CC=CO1 |
| C10H14O2 | |
| MFCD00037354 | |
| 2-hexanoylfuran, 2-furyl pentyl ketone, 1-hexanone, 1-2-furanyl, pentyl 2-furyl ketone, 1-2-furyl-1-hexanone, 1-furan-2-yl hexan-1-one, 1-2-furyl hexanone, 2-furyl n-pentyl ketone, 1-2-furanyl-1-hexanone, furan, 2-hexanoyl | |
| 1-(furan-2-yl)hexan-1-one |
Specifications
| 14360-50-0 | |
| 125°C to 127°C (15 mmHg) | |
| Characteristic | |
| 1.489 | |
| 5 g | |
| 2-hexanoylfuran, 2-furyl pentyl ketone, 1-hexanone, 1-2-furanyl, pentyl 2-furyl ketone, 1-2-furyl-1-hexanone, 1-furan-2-yl hexan-1-one, 1-2-furyl hexanone, 2-furyl n-pentyl ketone, 1-2-furanyl-1-hexanone, furan, 2-hexanoyl | |
| YUAYWSBSIJVIBS-UHFFFAOYSA-N | |
| 1-(furan-2-yl)hexan-1-one | |
| 61738 | |
| 99% |
| 0.994 | |
| 107°C (225°F) | |
| C10H14O2 | |
| MFCD00037354 | |
| 2956 | |
| Difficult to mix. | |
| CCCCCC(=O)C1=CC=CO1 | |
| 166.22 | |
| 166.22 | |
| 2-Hexanoylfuran |
Safety and Handling
EINECSNumber : 238-333-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only