missing translation for 'onlineSavingsMsg'
Learn More

2-(Hexamethyleneimino)ethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. p-7058261
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0183322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0183322 Supplier Thermo Scientific Chemicals Supplier No. L0183322
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26487-67-2
Molecular Formula C8H17Cl2N
Molecular Weight (g/mol) 198.131
MDL Number MFCD00012842
InChI Key ZQDSOUPBYJIPNM-UHFFFAOYSA-N
Synonym 1-2-chloroethyl azepane hydrochloride, 2-hexamethyleneimino ethyl chloride hydrochloride, 1h-azepine, hydrochloride, 2-chloroethylhexamethylenemine hydrochloride, 1-2-chloroethyl hexamethyleneimine hydrochloride, 2-chloroethyl hexamethylene imine hydrochloride, 2-chloroethyl azaperhydroepine, chloride, pubchem21389, 1-2-chloroethyl azepane hcl, wln: t7ntj a2g &gh
PubChem CID 24188283
IUPAC Name 1-(2-chloroethyl)azepane;hydrochloride
SMILES C1CCCN(CC1)CCCl.Cl

Specifications

Melting Point 206°C to 212°C
Quantity 100 g
UN Number UN2811
Beilstein 3680388
Sensitivity Hygroscopic
Formula Weight 198.14
Percent Purity 98%
Chemical Name or Material 2-(Hexamethyleneimino)ethyl chloride hydrochloride
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 247-733-4
RTECSNumber CM3185000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only