missing translation for 'onlineSavingsMsg'
Learn More

2-Hexadecyloctadecanoic Acid 98.0+%, TCI America™

Catalog No. H08371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H08371G Supplier TCI America Supplier No. H08371G
Only null left

Chemical Identifiers

CAS 89547-15-9
Molecular Formula C34H68O2
Molecular Weight (g/mol) 508.916
MDL Number MFCD00191519
InChI Key LPZKXVVBAZTEMK-UHFFFAOYSA-N
Synonym 2-Hexadecylstearic Acid
PubChem CID 625165
IUPAC Name 2-hexadecyloctadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(CCCCCCCCCCCCCCCC)C(=O)O

Specifications

Melting Point 79°C
Color White
Boiling Point 350°C
Quantity 1 g
Formula Weight 508.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hexadecyloctadecanoic Acid