missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyne 99.0+%, TCI America™

Catalog No. H04315ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04315ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04315ML Supplier TCI America Supplier No. H04315ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1119-65-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00039970
InChI Key AMSFEMSYKQQCHL-UHFFFAOYSA-N
Synonym 2-heptyne, 1-methyl-2-butylacetylene, butylmethylacetylene, n-butylmethylacetylene, acmc-1brzr, n-c4h9c.$.cch3, 2-heptyne,
PubChem CID 14245
IUPAC Name hept-2-yne
SMILES CCCCC#CC

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 mL
UN Number 3295
Formula Weight 96.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptyne