missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyne, 97%, Thermo Scientific™

Catalog No. p-7077563
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455560050 5g
AC455560250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455560050 Supplier Thermo Scientific Chemicals Supplier No. 455560050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1119-65-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
InChI Key AMSFEMSYKQQCHL-UHFFFAOYSA-N
Synonym 2-heptyne, 1-methyl-2-butylacetylene, butylmethylacetylene, n-butylmethylacetylene, acmc-1brzr, n-c4h9c.$.cch3, 2-heptyne,
PubChem CID 14245
IUPAC Name hept-2-yne
SMILES CCCCC#CC

Specifications

Density 0.7590g/mL
Boiling Point 110.0°C to 111.0°C (743.0 mmHg)
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3C≡CCH3
Quantity 5g
Specific Gravity 0.759
Formula Weight 96.17
Percent Purity 97%
Chemical Name or Material 2-Heptyne
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 214-285-6

RUO – Research Use Only