missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptyne, 97%, Thermo Scientific™
Specifications
Specifications
| Density | 0.7590g/mL |
| Boiling Point | 110.0°C to 111.0°C (743.0 mmHg) |
| Flash Point | 11°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Linear Formula | CH3(CH2)3C≡CCH3 |
| Quantity | 25g |
| Specific Gravity | 0.759 |
| Formula Weight | 96.17 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 214-285-6 |
RUO – Research Use Only