missing translation for 'onlineSavingsMsg'
Learn More

2-Heptylamine 97.0+%, TCI America™

Catalog No. p-7130656
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H064525ML 25 mL
H0645250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H064525ML Supplier TCI America Supplier No. H064525ML
Only null left

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 mL
UN Number 2734
Formula Weight 115.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Heptylamine
RTECSNumber MQ5425000
TSCA Yes