missing translation for 'onlineSavingsMsg'
Learn More

2-Heptylamine, 98+%

Catalog No. AC103770250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103770250 25 mL Glass bottle
AC103771000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103770250 Supplier Thermo Scientific Chemicals Supplier No. 103770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 123-82-0 | C7H17N | 115.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7680g/mL
Boiling Point 142.0°C to 144.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4175 to 1.4195
Linear Formula CH3(CH2)4CH(NH2)CH3
Quantity 25 mL
Beilstein 04,194
Merck Index 15,9982
Specific Gravity 0.768
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, ether, petroleum ether,, chloroform and benzene
Formula Weight 115.22
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Heptylamine, ≥98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rin
EINECSNumber 204-655-5
RTECSNumber MQ5425000
TSCA TSCA

RUO – Research Use Only