missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanol, 99+%

CAS: 543-49-7 | C7H16O | 116.20 g/mol

$68.13

Chemical Identifiers

CAS 543-49-7
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004587
InChI Key CETWDUZRCINIHU-UHFFFAOYNA-N
Synonym 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
PubChem CID 10976
IUPAC Name heptan-2-ol
SMILES CCCCCC(C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC120370500
View Documents Promotion Details
N/A 50 mL Glass bottle
Each for $68.13
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

543-49-7
116.20
CETWDUZRCINIHU-UHFFFAOYNA-N
10976
CCCCCC(C)O
C7H16O
MFCD00004587
2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
heptan-2-ol

Specifications

543-49-7
100.0
0.8175g/mL
59°C
99% min. (GC)
1.4200 to 1.422
MFCD00004587
0.8175
2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
CETWDUZRCINIHU-UHFFFAOYNA-N
heptan-2-ol
10976
99+%
2-Heptanol, 99+%
99.0
Colorless
160°C to 162°C
Authentic
C7H16O
CH3(CH2)4CH(OH)CH3
01, 415
15, 4697
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,benzene,miscible with most common organic liquids
CCCCCC(C)O
116.20
116.2
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or

GHS Signal Word: Warning

EINECSNumber : 208-844-3

RTECSNumber : MJ2975000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only