missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Heptanol, 98%

Catalog No. AAA1839118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1839118 50 g
AAA1839130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1839118 Supplier Thermo Scientific Chemicals Supplier No. A1839118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 543-49-7 | C7H16O | 116.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 543-49-7
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004587
InChI Key CETWDUZRCINIHU-UHFFFAOYNA-N
Synonym 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
PubChem CID 10976
IUPAC Name heptan-2-ol
SMILES CCCCCC(C)O

Specifications

Density 0.819
Boiling Point 160°C to 162°C
Flash Point 59°C (138°F)
Refractive Index 1.421
Quantity 50 g
UN Number UN1987
Beilstein 1719088
Merck Index 14,4662
Formula Weight 116.2
Percent Purity 98%
Chemical Name or Material (+/-)-2-Heptanol
Show More Show Less
Hazard Category H226-H312-H315-H319-H335
Hazard Statement GHS H Statement
H311-H226-H319
Toxic in contact with skin.
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 208-844-3
RTECSNumber MJ2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only