missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Heptafluoropropoxy)hexafluoropropyl Trifluorovinyl Ether 98.0+%, TCI America™
Supplier: TCI America P12265G
Specifications
| 2-(Heptafluoropropoxy)hexafluoropropyl Trifluorovinyl Ether | |
| Yellow | |
| C8F16O2 | |
| 5 g | |
| RJBJXVAPYONTFE-UHFFFAOYNA-N | |
| 1,1,1,2,2,3,3-heptafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propane | |
| 102662 | |
| ≥98.0% (GC) |
| 1644-11-7 | |
| 104°C | |
| MFCD00191479 | |
| 2-(Perfluoropropoxy)perfluoropropyl Trifluorovinyl Ether | |
| FC(F)=C(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F | |
| 432.06 | |
| 432.06 | |
| Liquid |
Chemical Identifiers
| 1644-11-7 | |
| 432.06 | |
| RJBJXVAPYONTFE-UHFFFAOYNA-N | |
| 102662 | |
| FC(F)=C(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F |
| C8F16O2 | |
| MFCD00191479 | |
| 2-(Perfluoropropoxy)perfluoropropyl Trifluorovinyl Ether | |
| 1,1,1,2,2,3,3-heptafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator