missing translation for 'onlineSavingsMsg'
Learn More

2-Furoylacetonitrile, 97%

Catalog No. AAA1449703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1449703 1 g
AAA1449706 5 g
AAA1449714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1449703 Supplier Thermo Scientific Chemicals Supplier No. A1449703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31909-58-7 | C7H5NO2 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31909-58-7
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00052210
InChI Key RZNSHBXVTAHWPP-UHFFFAOYSA-N
Synonym 2-furoylacetonitrile, 3-furan-2-yl-3-oxopropanenitrile, 3-2-furyl-3-oxopropanenitrile, 2-cyanoacetylfuran, 2-furanoylacetonitrile, 2-furoyl acetonitrile, pubchem11959, acmc-1cock, 2-2-furanoyl acetonitrile
PubChem CID 141671
IUPAC Name 3-(furan-2-yl)-3-oxopropanenitrile
SMILES O=C(CC#N)C1=CC=CO1

Specifications

Melting Point 77°C to 80°C
Quantity 1 g
Beilstein 111902
Formula Weight 135.12
Percent Purity 97%
Chemical Name or Material 2-Furoylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only