missing translation for 'onlineSavingsMsg'
Learn More

2-Furonitrile 98.0+%, TCI America™

Catalog No. p-7125103
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16485G 5 g
C164825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16485G Supplier TCI America Supplier No. C16485G

Chemical Identifiers

CAS 617-90-3
Molecular Formula C5H3NO
Molecular Weight (g/mol) 93.09
MDL Number MFCD00003223
InChI Key YXDXXGXWFJCXEB-UHFFFAOYSA-N
Synonym 2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile
PubChem CID 69245
IUPAC Name furan-2-carbonitrile
SMILES N#CC1=CC=CO1

Specifications

Boiling Point 148°C
Quantity 5 g
UN Number 1993
Formula Weight 93.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Furonitrile