missing translation for 'onlineSavingsMsg'
Learn More

2-Furonitrile, 98+%

Catalog No. p-7081988
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1221406 5 g
AAA1221414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1221406 Supplier Thermo Scientific Chemicals Supplier No. A1221406

CAS: 617-90-3 | C5H3NO | 93.09 g/mol

2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.

Solubility
Soluble in most common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 617-90-3
Molecular Formula C5H3NO
Molecular Weight (g/mol) 93.09
MDL Number MFCD00003223
InChI Key YXDXXGXWFJCXEB-UHFFFAOYSA-N
Synonym 2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile
PubChem CID 69245
IUPAC Name furan-2-carbonitrile
SMILES N#CC1=CC=CO1

Specifications

Density 1.062
Boiling Point 146°C to 148°C
Flash Point 35°C (95°F)
Odor Recognizable
Refractive Index 1.479
Quantity 5 g
UN Number UN1992
Beilstein 107033
Solubility Information Soluble in most common organic solvents. Slightly soluble in water.
Formula Weight 93.09
Percent Purity ≥98%
Chemical Name or Material 2-Furonitrile
Show More Show Less
missing translation for 'healthHazard1' H226-H302+H312+H332-H318
missing translation for 'healthHazard2' GHS H Statement
H311-H226-H302-H332
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
missing translation for 'healthHazard3' P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 210-537-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only