missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid hydrazide, 98%

Catalog No. AAA1363018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1363018 50 g
AAA1363009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1363018 Supplier Thermo Scientific Chemicals Supplier No. A1363018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3326-71-4 | C5H6N2O2 | 126.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3326-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00003235
InChI Key SKTSVWWOAIAIKI-UHFFFAOYSA-N
Synonym 2-furoic acid hydrazide, 2-furoic hydrazide, 2-furohydrazide, furoylhydrazide, 2-furancarbohydrazide, 2-furoylhydrazide, furoic acid, hydrazide, 2-furoic acid, hydrazide, 2-furoylhydrazine, 2-furylcarbonylhydrazide
PubChem CID 18731
IUPAC Name furan-2-carbohydrazide
SMILES C1=COC(=C1)C(=O)NN

Specifications

Melting Point 77°C to 81°C
Quantity 50 g
Beilstein 114435
Formula Weight 126.12
Percent Purity 98%
Chemical Name or Material 2-Furoic acid hydrazide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 222-046-2
RTECSNumber LV1925000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only