missing translation for 'onlineSavingsMsg'
Learn More

2-Furamide, 97%

Catalog No. AAB2507514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2507514 25 g
AAB2507503 1 g
AAB2507506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2507514 Supplier Thermo Scientific Chemicals Supplier No. B2507514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 609-38-1 | C5H5NO2 | 111.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-38-1
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00236147
InChI Key TVFIYRKPCACCNL-UHFFFAOYSA-N
Synonym 2-furamide, furamide, 2-furancarboxamide, furfurylamide, 2-furoamide, furoylamide, unii-8ig4632c1y, furancarboxamide, acmc-20anu6
PubChem CID 69108
IUPAC Name furan-2-carboxamide
SMILES C1=COC(=C1)C(=O)N

Specifications

Melting Point 139°C to 142°C
Quantity 25 g
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2-Furamide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only