missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Furaldehyde Diethyl Acetal 98.0+%, TCI America™
$76.43 - $188.27
Chemical Identifiers
| CAS | 13529-27-6 |
|---|---|
| Molecular Formula | C9H14O3 |
| Molecular Weight (g/mol) | 170.21 |
| MDL Number | MFCD00003245 |
| InChI Key | SEILDMUKBMYIEZ-UHFFFAOYSA-N |
| Synonym | 2-diethoxymethyl furan, 2-furaldehyde diethyl acetal, furfural diethyl acetal, furan, 2-diethoxymethyl, unii-04b3gme3ij, 2-furaldehydediethylacetal, 2-diethoxymethyl-furan, furan, diethoxymethyl, 2-furaldehyde diethylacetal, 04b3gme3ij |
| PubChem CID | 83553 |
| ChEBI | CHEBI:34769 |
| IUPAC Name | 2-(diethoxymethyl)furan |
| SMILES | CCOC(OCC)C1=CC=CO1 |
Chemical Identifiers
| 13529-27-6 | |
| 170.21 | |
| SEILDMUKBMYIEZ-UHFFFAOYSA-N | |
| 83553 | |
| 2-(diethoxymethyl)furan |
| C9H14O3 | |
| MFCD00003245 | |
| 2-diethoxymethyl furan, 2-furaldehyde diethyl acetal, furfural diethyl acetal, furan, 2-diethoxymethyl, unii-04b3gme3ij, 2-furaldehydediethylacetal, 2-diethoxymethyl-furan, furan, diethoxymethyl, 2-furaldehyde diethylacetal, 04b3gme3ij | |
| CHEBI:34769 | |
| CCOC(OCC)C1=CC=CO1 |
Specifications
| 13529-27-6 | |
| 81°C | |
| MFCD00003245 | |
| 2-diethoxymethyl furan, 2-furaldehyde diethyl acetal, furfural diethyl acetal, furan, 2-diethoxymethyl, unii-04b3gme3ij, 2-furaldehydediethylacetal, 2-diethoxymethyl-furan, furan, diethoxymethyl, 2-furaldehyde diethylacetal, 04b3gme3ij | |
| CCOC(OCC)C1=CC=CO1 | |
| 170.21 | |
| CHEBI:34769 | |
| ≥98.0% (GC) | |
| 2-Furaldehyde Diethyl Acetal |
| Yellow | |
| C9H14O3 | |
| 5 g | |
| SEILDMUKBMYIEZ-UHFFFAOYSA-N | |
| 2-(diethoxymethyl)furan | |
| 83553 | |
| 170.21 | |
| Liquid |
Safety and Handling
RTECSNumber : LU0585000
TSCA : Yes