missing translation for 'onlineSavingsMsg'
Learn More

2-Formylthiophene-4-boronic acid, 98%, Thermo Scientific™

$165.41 - $165.41

Chemical Identifiers

CAS 175592-59-3
Molecular Formula C5H5BO3S
Molecular Weight (g/mol) 155.962
MDL Number MFCD03002365
InChI Key ZCUFJAORCNFWOF-UHFFFAOYSA-N
Synonym 2-formylthiophene-4-boronic acid, 5-formylthiophen-3-yl boronic acid, 5-formylthiophene-3-boronic acid, 5-formyl-3-thienyl boronic acid, 4-borono-2-formylthiophene, 5-formylthiophen-3-boronic acid, 5-formyl-3-thienylboronic acid, 5-formyl-3-thiopheneboronic acid, 4-boronothiophene-2-carboxaldehyde, 2-formyl-4-thiopheneboronic acid
PubChem CID 11084115
IUPAC Name (5-formylthiophen-3-yl)boronic acid
SMILES B(C1=CSC(=C1)C=O)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAH5329803
View Documents
Thermo Scientific Chemicals
H5329803
1 g
Each for $165.41
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Formylthiophene-4-boronic acid is used as intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat. Refrigerate.
Specifications

Chemical Identifiers

175592-59-3
155.962
ZCUFJAORCNFWOF-UHFFFAOYSA-N
11084115
B(C1=CSC(=C1)C=O)(O)O
C5H5BO3S
MFCD03002365
2-formylthiophene-4-boronic acid, 5-formylthiophen-3-yl boronic acid, 5-formylthiophene-3-boronic acid, 5-formyl-3-thienyl boronic acid, 4-borono-2-formylthiophene, 5-formylthiophen-3-boronic acid, 5-formyl-3-thienylboronic acid, 5-formyl-3-thiopheneboronic acid, 4-boronothiophene-2-carboxaldehyde, 2-formyl-4-thiopheneboronic acid
(5-formylthiophen-3-yl)boronic acid

Specifications

175592-59-3
C5H5BO3S
1 g
Slightly soluble in water.
B(C1=CSC(=C1)C=O)(O)O
155.962
156.97
2-Formylthiophene-4-boronic acid
113°C
MFCD03002365
2-formylthiophene-4-boronic acid, 5-formylthiophen-3-yl boronic acid, 5-formylthiophene-3-boronic acid, 5-formyl-3-thienyl boronic acid, 4-borono-2-formylthiophene, 5-formylthiophen-3-boronic acid, 5-formyl-3-thienylboronic acid, 5-formyl-3-thiopheneboronic acid, 4-boronothiophene-2-carboxaldehyde, 2-formyl-4-thiopheneboronic acid
ZCUFJAORCNFWOF-UHFFFAOYSA-N
(5-formylthiophen-3-yl)boronic acid
11084115
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents