missing translation for 'onlineSavingsMsg'
Learn More

2-Formylselenophene (stabilized with HQ) 98.0+%, TCI America™

Catalog No. p-7129896
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07821G 1 g
F07825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07821G Supplier TCI America Supplier No. F07821G
Only null left

Chemical Identifiers

CAS 25109-26-6
Molecular Formula C5H4OSe
Molecular Weight (g/mol) 159.057
InChI Key AGBORNIRMFZCGC-UHFFFAOYSA-N
Synonym 2-Selenophenecarboxaldehyde
PubChem CID 141174
IUPAC Name selenophene-2-carbaldehyde
SMILES C1=C[Se]C(=C1)C=O

Specifications

Color Red-Yellow
Boiling Point 85°C
Quantity 1 g
UN Number 3440
Formula Weight 159.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Formylselenophene (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator