missing translation for 'onlineSavingsMsg'
Learn More

2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH6221114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6221114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6221114 Supplier Thermo Scientific Chemicals Supplier No. H6221114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 94744-50-0 | Molecular Formula: C19H19NO4 | Molecular Weight: 325.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94744-50-0
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.36
MDL Number MFCD00151913
InChI Key HOZZVEPRYYCBTO-UHFFFAOYSA-N
Synonym fmoc-aib-oh, fmoc-alpha-methylalanine, fmoc-alpha-me-ala-oh, 2-fmoc-amino isobutyric acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-2-methylpropanoic acid, alanine, n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methylpropanoic acid, n-fmoc-2-aminoisobutyric acid, 2-9h-fluoren-9-ylmethoxy carbonylamino isobutyric acid
PubChem CID 2756096
SMILES CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 182°C to 188°C
Quantity 25 g
Beilstein 5604328
Sensitivity Moisture sensitive
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-methylpropanoic acid
Formula Weight 325.36
Percent Purity 98%
Chemical Name or Material 2-(Fmoc-amino)isobutyric acid
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only