missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine 98.0+%, TCI America™

Supplier:  TCI America F0217100G

 View more versions of this product

Catalog No. F0217100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Fluoropyridine
Yellow
C5H4FN
100 g
pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
C1=CC=NC(=C1)F
97.092
97.09
Liquid
372-48-5
126°C
MFCD00006224
1993
MTAODLNXWYIKSO-UHFFFAOYSA-N
2-fluoropyridine
9746
≥98.0% (GC)

Chemical Identifiers

372-48-5
97.092
MTAODLNXWYIKSO-UHFFFAOYSA-N
9746
C1=CC=NC(=C1)F
C5H4FN
MFCD00006224
pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
2-fluoropyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions