missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine 98.0+%, TCI America™

Catalog No. F021725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F021725G 25 g
F0217100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F021725G Supplier TCI America Supplier No. F021725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372-48-5 | C5H4FN | 97.092 g/mol

Chemical Identifiers

CAS 372-48-5
Molecular Formula C5H4FN
Molecular Weight (g/mol) 97.092
MDL Number MFCD00006224
InChI Key MTAODLNXWYIKSO-UHFFFAOYSA-N
Synonym pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
PubChem CID 9746
IUPAC Name 2-fluoropyridine
SMILES C1=CC=NC(=C1)F

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 1993
Formula Weight 97.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoropyridine