Learn More
2-Fluoropyridine, 99%
CAS: 372-48-5 | C5H4FN | 97.092 g/mol
Supplier: Thermo Scientific Chemicals A1162714
Description
2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.
Solubility
Soluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
| 2-Fluoropyridine | |
| 1.129 | |
| 24°C (75°F) | |
| 1.467 | |
| 25 g | |
| 1515 | |
| Soluble in water. | |
| C1=CC=NC(=C1)F | |
| 97.092 | |
| 97.09 |
| 372-48-5 | |
| 124°C to 126°C | |
| C5H4FN | |
| MFCD00006224 | |
| UN1993 | |
| pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium | |
| MTAODLNXWYIKSO-UHFFFAOYSA-N | |
| 2-fluoropyridine | |
| 9746 | |
| 99% |
Chemical Identifiers
| 372-48-5 | |
| 97.092 | |
| MTAODLNXWYIKSO-UHFFFAOYSA-N | |
| 9746 | |
| C1=CC=NC(=C1)F |
| C5H4FN | |
| MFCD00006224 | |
| pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium | |
| 2-fluoropyridine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-757-5
RTECSNumber : UT3900000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only