missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoropyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™
$78.93 - $78.93
Specifications
| CAS | 351019-18-6 |
|---|---|
| Melting Point | 172°C |
| Color | White-Yellow |
| Molecular Formula | C5H5BFNO2 |
| MDL Number | MFCD03411559 |
Specifications
| 351019-18-6 | |
| White-Yellow | |
| MFCD03411559 | |
| (6-fluoropyridin-3-yl)boronic acid | |
| 140.91 |
| 172°C | |
| C5H5BFNO2 | |
| OJBYZWHAPXIJID-UHFFFAOYSA-N | |
| 2783397 | |
| Crystalline Powder |