missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F07701G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07701G Supplier TCI America Supplier No. F07701G

Specifications

Chemical Name or Material 2-Fluoropyridine-5-boronic Acid (contains varying amounts of Anhydride)
CAS 351019-18-6
Melting Point 172°C
Color White-Yellow
Molecular Formula C5H5BFNO2
Quantity 1 g
MDL Number MFCD03411559
Synonym 2-fluoropyridine-5-boronic acid, 2-fluoro-5-pyridylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridine-3-boronic acid, 2-fluoro-5-pyridineboronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridin-5-boronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoro-3-pyridineboronic acid
InChI Key OJBYZWHAPXIJID-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(F)N=C1
IUPAC Name (6-fluoropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 140.91
PubChem CID 2783397
Formula Weight 140.91
Physical Form Crystalline Powder
Show More Show Less