missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
$347.77 - $347.77
Chemical Identifiers
| CAS | 402-65-3 |
|---|---|
| Molecular Formula | C6H4FNO2 |
| Molecular Weight (g/mol) | 141.10 |
| MDL Number | MFCD02181194 |
| InChI Key | JMPFWDWYGOWUFP-UHFFFAOYSA-N |
| Synonym | 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548 |
| PubChem CID | 345360 |
| IUPAC Name | 2-fluoropyridine-4-carboxylic acid |
| SMILES | OC(=O)C1=CC=NC(F)=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 402-65-3 | |
| 141.10 | |
| JMPFWDWYGOWUFP-UHFFFAOYSA-N | |
| 345360 | |
| OC(=O)C1=CC=NC(F)=C1 |
| C6H4FNO2 | |
| MFCD02181194 | |
| 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548 | |
| 2-fluoropyridine-4-carboxylic acid |
Specifications
| 402-65-3 | |
| C6H4FNO2 | |
| 1 g | |
| JMPFWDWYGOWUFP-UHFFFAOYSA-N | |
| 2-fluoropyridine-4-carboxylic acid | |
| 345360 | |
| 98% | |
| 2-Fluoropyridine-4-carboxylic acid |
| 195°C to 497°C | |
| MFCD02181194 | |
| 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548 | |
| OC(=O)C1=CC=NC(F)=C1 | |
| 141.10 | |
| 141.1 | |
| Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only