missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F09351G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09351G Supplier TCI America Supplier No. F09351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 401815-98-3 | C5H5BFNO2 | 140.91 g/mol

Specifications

Chemical Name or Material 2-Fluoropyridine-4-boronic Acid (contains varying amounts of Anhydride)
CAS 401815-98-3
Melting Point 200°C
Color White
Molecular Formula C5H5BFNO2
MDL Number MFCD04112534
Quantity 1 g
Synonym 2-fluoropyridine-4-boronic acid, 2-fluoropyridin-4-yl boronic acid, 2-fluoro-4-pyridineboronic acid, 2-fluoro-4-pyridinylboronic acid, 2-fluoropyridine-4-boronicacid, 2-fluoropyridin-4-yl-4-boronic acid, 2-fluoropyridine-boronic acid, 2-fluoro-4-pyridyl boronic acid, 2-fluoro-pyridine-4-boronic acid, boronic acid, 2-fluoro-4-pyridinyl
InChI Key WXGBZJJAGLSBPR-UHFFFAOYSA-N
SMILES OB(O)C1=CC=NC(F)=C1
IUPAC Name (2-fluoropyridin-4-yl)boronic acid
Molecular Weight (g/mol) 140.91
PubChem CID 2783396
Formula Weight 140.91
Percent Purity 98.0 to 115.0%
Physical Form Crystalline Powder
Show More Show Less