missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH2763006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2763006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2763006 Supplier Thermo Scientific Chemicals Supplier No. H2763006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 401815-98-3 | Molecular Formula: C5H5BFNO2 | Molecular Weight: 140.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401815-98-3
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
MDL Number MFCD04112534
InChI Key WXGBZJJAGLSBPR-UHFFFAOYSA-N
Synonym 2-fluoropyridine-4-boronic acid, 2-fluoropyridin-4-yl boronic acid, 2-fluoro-4-pyridineboronic acid, 2-fluoro-4-pyridinylboronic acid, 2-fluoropyridine-4-boronicacid, 2-fluoropyridin-4-yl-4-boronic acid, 2-fluoropyridine-boronic acid, 2-fluoro-4-pyridyl boronic acid, 2-fluoro-pyridine-4-boronic acid, boronic acid, 2-fluoro-4-pyridinyl
PubChem CID 2783396
IUPAC Name (2-fluoropyridin-4-yl)boronic acid
SMILES OB(O)C1=CC=NC(F)=C1

Specifications

Melting Point ∽195°C (decomposition)
Quantity 5 g
Formula Weight 140.91
Percent Purity 95%
Chemical Name or Material 2-Fluoropyridine-4-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only