missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Fluoropropiophenone 98.0+%, TCI America™
Supplier: TCI America F045025G
Specifications
| 2′-Fluoropropiophenone | |
| Yellow | |
| MFCD00009893 | |
| 2'-fluoropropiophenone, 1-2-fluorophenyl propan-1-one, 1-2-fluorophenyl-1-propanone, ethyl 2-fluorophenyl ketone, 1-propanone, 1-2-fluorophenyl, 1-2-fluorphenyl propan-1-on, 2;-fluoropropiophenone, pubchem19917, acmc-1cpyw, o-fluoropropiophenone | |
| CCC(=O)C1=CC=CC=C1F | |
| 152.17 | |
| 152.17 | |
| Liquid |
| 446-22-0 | |
| C9H9FO | |
| 25 g | |
| NSNSIFGTEGKZFK-UHFFFAOYSA-N | |
| 1-(2-fluorophenyl)propan-1-one | |
| 579399 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 446-22-0 | |
| 152.17 | |
| NSNSIFGTEGKZFK-UHFFFAOYSA-N | |
| 579399 | |
| CCC(=O)C1=CC=CC=C1F |
| C9H9FO | |
| MFCD00009893 | |
| 2'-fluoropropiophenone, 1-2-fluorophenyl propan-1-one, 1-2-fluorophenyl-1-propanone, ethyl 2-fluorophenyl ketone, 1-propanone, 1-2-fluorophenyl, 1-2-fluorphenyl propan-1-on, 2;-fluoropropiophenone, pubchem19917, acmc-1cpyw, o-fluoropropiophenone | |
| 1-(2-fluorophenyl)propan-1-one |
Safety and Handling
TSCA : No