missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoropropiophenone 98.0+%, TCI America™

Catalog No. F04505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04505G 5 g
F045025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04505G Supplier TCI America Supplier No. F04505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-22-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00009893
InChI Key NSNSIFGTEGKZFK-UHFFFAOYSA-N
Synonym 2'-fluoropropiophenone, 1-2-fluorophenyl propan-1-one, 1-2-fluorophenyl-1-propanone, ethyl 2-fluorophenyl ketone, 1-propanone, 1-2-fluorophenyl, 1-2-fluorphenyl propan-1-on, 2;-fluoropropiophenone, pubchem19917, acmc-1cpyw, o-fluoropropiophenone
PubChem CID 579399
IUPAC Name 1-(2-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=CC=C1F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 152.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Fluoropropiophenone