missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoropropiophenone, 99%

Catalog No. AAL1242304
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1242304 2 g
AAL1242309 10 g
AAL1242318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1242304 Supplier Thermo Scientific Chemicals Supplier No. L1242304
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 446-22-0 | C9H9FO | 152.17 g/mol

2'-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, methanol. Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 446-22-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00009893
InChI Key NSNSIFGTEGKZFK-UHFFFAOYSA-N
Synonym 2'-fluoropropiophenone, 1-2-fluorophenyl propan-1-one, 1-2-fluorophenyl-1-propanone, ethyl 2-fluorophenyl ketone, 1-propanone, 1-2-fluorophenyl, 1-2-fluorphenyl propan-1-on, 2;-fluoropropiophenone, pubchem19917, acmc-1cpyw, o-fluoropropiophenone
PubChem CID 579399
IUPAC Name 1-(2-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=CC=C1F

Specifications

Density 1.102
Boiling Point 89°C to 91°C (12 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.5045
Quantity 2 g
Beilstein 3235723
Solubility Information Soluble in chloroform,methanol. Not miscible in water.
Formula Weight 152.17
Percent Purity 99%
Chemical Name or Material 2'-Fluoropropiophenone
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only