missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenethyl bromide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 91319-54-9
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
InChI Key FQGDFWIQBCQXPS-UHFFFAOYSA-N
Synonym 2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
PubChem CID 2756964
IUPAC Name 1-(2-bromoethyl)-2-fluorobenzene
SMILES C1=CC=C(C(=C1)CCBr)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440120010
SDS View Documents
Thermo Scientific Chemicals
440120010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440120050
SDS View Documents
Thermo Scientific Chemicals
440120050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

91319-54-9
203.05
2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
1-(2-bromoethyl)-2-fluorobenzene
C8H8BrF
FQGDFWIQBCQXPS-UHFFFAOYSA-N
2756964
C1=CC=C(C(=C1)CCBr)F

Specifications

91319-54-9
100.0
Authentic
Glass Bottle
1g
FQGDFWIQBCQXPS-UHFFFAOYSA-N
1-(2-bromoethyl)-2-fluorobenzene
2756964
97%
96.0
73°C to 75°C (20.0 Torr)
96% min. (GC)
C8H8BrF
2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
C1=CC=C(C(=C1)CCBr)F
203.05
203.05
2-Fluorophenethyl bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only