missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoroisonicotinic Acid 97.0+%, TCI America™
$29.83 - $117.43
Chemical Identifiers
| CAS | 402-65-3 |
|---|---|
| Molecular Formula | C6H4FNO2 |
| Molecular Weight (g/mol) | 141.10 |
| MDL Number | MFCD02181194 |
| InChI Key | JMPFWDWYGOWUFP-UHFFFAOYSA-N |
| Synonym | 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548 |
| PubChem CID | 345360 |
| IUPAC Name | 2-fluoropyridine-4-carboxylic acid |
| SMILES | OC(=O)C1=CC=NC(F)=C1 |
Chemical Identifiers
| 402-65-3 | |
| 141.10 | |
| JMPFWDWYGOWUFP-UHFFFAOYSA-N | |
| 345360 | |
| OC(=O)C1=CC=NC(F)=C1 |
| C6H4FNO2 | |
| MFCD02181194 | |
| 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548 | |
| 2-fluoropyridine-4-carboxylic acid |
Specifications
| 402-65-3 | |
| White | |
| MFCD02181194 | |
| 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548 | |
| OC(=O)C1=CC=NC(F)=C1 | |
| 141.10 | |
| 141.10 | |
| Crystalline Powder |
| 191°C | |
| C6H4FNO2 | |
| 1 g | |
| JMPFWDWYGOWUFP-UHFFFAOYSA-N | |
| 2-fluoropyridine-4-carboxylic acid | |
| 345360 | |
| ≥97.0% (GC,T) | |
| 2-Fluoroisonicotinic Acid |
Safety and Handling
TSCA : No