missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroisonicotinic Acid 97.0+%, TCI America™

Catalog No. F06935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06935G 5 g
F06931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06935G Supplier TCI America Supplier No. F06935G

Chemical Identifiers

CAS 402-65-3
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD02181194
InChI Key JMPFWDWYGOWUFP-UHFFFAOYSA-N
Synonym 2-fluoroisonicotinic acid, 2-fluoro-4-pyridinecarboxylic acid, 2-fluoro-isonicotinic acid, 4-pyridinecarboxylic acid, 2-fluoro, 2-fluoroisonicotinicacid, 2-fluoro-4-carboxypyridine, 2-fluoro-4-pyridinylcarboxylic acid, 2-fluoropyridine-4-carboxylicacid, pubchem2998, pubchem3548
PubChem CID 345360
IUPAC Name 2-fluoropyridine-4-carboxylic acid
SMILES OC(=O)C1=CC=NC(F)=C1

Specifications

Melting Point 191°C
Color White
Quantity 5 g
Formula Weight 141.10
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoroisonicotinic Acid