missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorocinnamic acid, predominantly trans, 98%

Catalog No. p-7022489
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1530306 5 g
AAA1530314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1530306 Supplier Thermo Scientific Chemicals Supplier No. A1530306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 451-69-4 | C9H7FO2 | 166.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 451-69-4
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.15
MDL Number MFCD00004370
InChI Key IOUDZAFBPDDAMK-AATRIKPKSA-N
Synonym 2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid
PubChem CID 735833
IUPAC Name (E)-3-(2-fluorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1F

Specifications

Melting Point 179°C to 183°C
Odor Odorless
Quantity 5 g
Beilstein 1863963
Formula Weight 166.15
Percent Purity ≥98%
Chemical Name or Material 2-Fluorocinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-195-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only