missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorocinnamic acid, 98%, Thermo Scientific™

Numéro de catalogue. p-134050
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
25g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC207100010 1g Glass bottle
AC207100050 5g Glass bottle
AC207100250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC207100010 Fournisseur Thermo Scientific Chemicals Code fournisseur 207100010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 451-69-4
Formule moléculaire C9H7FO2
Poids moléculaire (g/mol) 166.15
Numéro MDL MFCD00004370
Clé InChI IOUDZAFBPDDAMK-AATRIKPKSA-N
Synonyme 2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid
PubChem CID 735833
Nom de l’IUPAC (E)-3-(2-fluorophenyl)prop-2-enoic acid
SOURIRES OC(=O)\C=C\C1=CC=CC=C1F

Spécifications

Nom ou substance chimique 2-Fluorocinnamic acid
Nom Remarque 98%
CAS % minimum 97.5
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98%
Emballage Glass bottle
Formule linéaire FC6H4CH=CHCO2H
Quantité 1g
Beilstein 09, 594
Poids de formule 166.15
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-195-3
RTECSNumber GD9081000
TSCA TSCA

RUO – Research Use Only