missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzothiazole, 99%

Catalog No. p-7047489
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19316MD 250 mg
AAL1931603 1 g
AAL1931606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19316MD Supplier Thermo Scientific Chemicals Supplier No. L19316MD

CAS: 1123-98-4 | C7H4FNS | 153.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1123-98-4
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00047254
InChI Key QVWCHVAUHZEAAT-UHFFFAOYSA-N
Synonym 2-fluorobenzothiazole, 2-fluorobenzo d thiazole, 2-benzothiazolyl fluoride, benzothiazole, 2-fluoro, 2-fluoro-benzothiazole, fluorobenzothiazole, acmc-2099en, #
PubChem CID 70753
IUPAC Name 2-fluoro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)F

Specifications

Density 1.33
Boiling Point 97°C to 98°C (16 mmHg)
Odor Characteristic
Refractive Index 1.583
Quantity 250 mg
UN Number UN2810
Beilstein 971503
Formula Weight 153.18
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzothiazole
Hazard Category H227-H301+H311+H331
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 214-384-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only